Anti-SYF2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-661EA 420 EUR
PRSI45-661
Anti-SYF2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 26 kDa band seen in mouse heart lysate and 30 kDa band seen in human heart lysate (Predicted MW of 28 kDa according to NP_056299). Recommended for use at 0.5-3 μg/mL.

GCIP Interacting Protein antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GCIP Interacting Protein antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SYF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR