Anti-CCNDBP1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-660EA 553 EUR
PRSI45-660
Anti-CCNDBP1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution >1/32,000. Western Blot: Approximately 55 kDa band in K562 lysate at 0.5 μg/mL. This band was successfully blocked by incubation with the immunizing peptide. Immunohistochemistry: In paraffin embedded human prostate shows textured cytoplasm staining with emphasis on the luminal side of the secretory cells in the glands. Recommended concentration, 3-6 μg/mL.

GCIP antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GCIP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CCNDBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR