Anti-GALR1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-653EA 553 EUR
PRSI45-653
Anti-GALR1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 24-25 kDa and 37 kDa bands observed in lysates of rat spinal cord (calculated MW of 24.9 kDa according to rat NP_037090.1 and calculated MW of 38.9 kDa according to AAC52438.1 and sp Q62805). Recommended concentration: 0.1-0.3 μg/mL.

GALR1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GALR1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GALR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR