Anti-GAD2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-648EA 420 EUR
PRSI45-648
Anti-GAD2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 65+70 kDa bands observed in human brain lysates. an additional band of 26 kDa is observed. All bands were blocked by the immunizing peptide (calculated MW of 65.4 kDa according to NP_000809.1). Recommended concentration: 0.1-0.3 μg/mL.

GAD65 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GAD65 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GAD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR