Anti-GABPB2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-647EA 345 EUR
PRSI45-647
Anti-GABPB2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 35 kDa band in rat kidney lysates after 1 μg/mL antibody staining.7 kDa according to NP_766100.1 and NP_084161.1. The 35 kDa band was successfully blocked by incubation with the immunizing peptide.

GABPB2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GABPB2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Gabpb2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR