Anti-GABPB2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-646EA 420 EUR
PRSI45-646
Anti-GABPB2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 45+38 kDa bands observed in HeLa lysate (calculated MW of 42.5 kDa according to NP_005245.2 and of 38.1 kDa according to NP_002032.2). Recommended concentration: 0.5-1.5 μg/mL. an additional band of unknown identity was also consistently observed at 21 kDa . This band was successfully blocked by incubation with the immunizing peptide.

GABPB2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GABPB2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GABPB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR