Anti-FH Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-641EA 420 EUR
PRSI45-641
Anti-FH Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Approximately 48 kDa band observed in human kidney, liver and lung lysates (calculated MW of 54.6 kDa according to NP_000134.2). The observed molecular weight corresponds to earlier findings with antibodies from different sources. In transfected HEK293 transiently expressing Fumarase a band of approximately 55 kDa is observed. This band is not observed in the non-transfected HEK293.Recommended concentration: 0.01-0.03 μg/mL.

Fumarase antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Fumarase antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR