Anti-FZD8 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-636EA 420 EUR
PRSI45-636
Anti-FZD8 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 75 kDa band observed in human heart and human pancreas lysates (calculated MW of 73.3 kDa according to NP_114072.1). Recommended concentration: 1-3 μg/mL. Immunohistochemistry: In paraffin embedded human pancreas, shows granular cytoplasmic staining of acinar cells. Cells of the Islets of Langerhans are weakly stained, if at all. Recommended concentration, 3 μg/mL.

Frizzled 8 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Frizzled 8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FZD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR