Anti-FOXE1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-608EA 553 EUR
PRSI45-608
Anti-FOXE1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 70 kDa band in human thymus and thyroid gland lysates after 0.3 μg/mL antibody staining.1 kDa according to NP_004464.2. The 65 kDa band was successfully blocked by incubation with the immunizing peptide. Immunohistochemistry: In paraffin embedded human testis shows strong nuclear staining in developing sperm cells. Recommended concentration, 2-3 μg/mL.

FOXE1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FOXE1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOXE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR