Anti-FBXW2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-583EA 553 EUR
PRSI45-583
Anti-FBXW2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 48 kDa band observed in human kidney lysates (calculated MW of 48.0 kDa according to NP_036296.1). Recommended concentration: 0.3-1.5 μg/mL. A stronger band of unknown identity was also consistently observed at appr. 30 kDa - both bands were successfully blocked by incubation with the immunizing peptide. Immunohistochemistry: In paraffin embedded human kidney, shows cytoplasmic staining of epithelial cells in some renal tubules. Recommended concentration, 2-3 μg/mL.

FBXW2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

FBXW2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FBXW2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR