Anti-UBE2O Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-512EA 553 EUR
PRSI45-512
Anti-UBE2O Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Immunohistochemistry: In paraffin embedded human heart shows textured staining in some myocardial fibres. Recommended concentration, 4-6 μg/mL.

E2-230K antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

E2-230K antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: UBE2O
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR