Anti-DRD5 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-497EA 345 EUR
PRSI45-497
Anti-DRD5 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 75-80 kDa band in lysates of human brain (hippocampus), kidney, spleen and thymus after 0.1 μg/mL antibody staining.0 kDa according to NP_000789.1. The 75-80 kDa band was successfully blocked by incubation with the immunizing peptide.

DRD5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

DRD5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DRD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR