Anti-DIAPH3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-478EA 345 EUR
PRSI45-478
Anti-DIAPH3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 50 kDa band in human bone marrow, liver and lymph node lysates after 0.3 μg/mL antibody staining.1 and 98. The ~50 kDa band was successfully blocked by incubation with the immunizing peptide.

DIAPH3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

DIApH 3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DIAPH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR