Anti-DCDC2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-464EA 420 EUR
PRSI45-464
Anti-DCDC2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: full length recombinant human DCDC2 expressed in COS7 (Lane A), non-transfectant COS7 (lane B). Approximately 52 kDa band observed (calculated MW of 52.8 kDa according to NP_057440.1). Recommmended concentration 0.1-0.3 μg/mL.

DCDC2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

DCDC2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DCDC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR