Anti-DAZL Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-462EA 420 EUR
PRSI45-462
Anti-DAZL Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 35 kDa band observed in human ovary and human testes lysates (calculated MW of 33.2 kDa according to NP_001342.2). Recommended concentration: 0.1-0.3 μg/mL. Immunohistochemistry: In paraffin embedded mouse testis shows cytoplasm staining of spermatogonia. Additional staining is observed in the cytoplasm of elongatng spermatids but this additional staining is not in agreement with results obtained by another antibody. Recommended concentration, 1-3 μg/mL.

DAZL antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

DAZL antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DAZL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR