Anti-HCST Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-460EA 420 EUR
PRSI45-460
Anti-HCST Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Approximately 40 kDa band observed in human spleen lysates (calculated MW of 9.5 kDa according to NP_055081.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Wu et al, Science. 1999 Jul 30;285 (5428) :730-2.; PMID: 10426994). Recommended concentration: 0.2-0.6 μg/mL. an additional band of unknown identity was also consistently observed at 100 kDa . This band was successfully blocked by incubation with the immunizing peptide.

DAP10 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

DAP10 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HCST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR