Anti-TPD52L1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-454EA 553 EUR
PRSI45-454
Anti-TPD52L1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 26 kDa band observed in human breast tumor lysates (calculated MW of 22.5 kDa according to NP_003278.1). Recommended concentration: 0.1-0.3 μg/mL. See ProSci's D52 and D54 products.

D53 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

D53 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TPD52L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR