Anti-CYBRD1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-452EA 553 EUR
PRSI45-452
Anti-CYBRD1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: experiments gave bands at approximately 35 kDa and a 26+28 kDa doublet in human colon lysate after 0.1 μg/mL antibody staining. The detected bands were all successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands). Immunohistochemistry: In paraffin embedded human testis shows membranous staining in primarily myofibroblasts of the basement membrane surrounding the seminiferous tubules. Recommended concentration, 2-5 μg/mL.

Cytochrome B Reductase 1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Cytochrome B Reductase 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CYBRD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR