Anti-CYP7B1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-449EA 420 EUR
PRSI45-449
Anti-CYP7B1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Preliminary experiments in human liver, kidney and prostate lysates gave no specific signal but low background (at antibody concentration up to 1 μg/mL).Immunohistochemistry: In paraffin embedded human breast shows textured cytoplasm staining in ductal epithelial cells. Recommended concentration, 3-6 μg/mL.

CYP7B1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CYP7B1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CYP7B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR