Anti-CXCR3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-439EA 553 EUR
PRSI45-439
Anti-CXCR3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 50-55 kDa band in lysates of human thymus, kidney and placenta, and of cell line HeLa after 0.3 μg/mL antibody staining. The 50-55 kDa band was successfully blocked by incubation with the immunizing peptide.

CXCR3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CXCR3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CXCR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR