Anti-CORO1A Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-421EA 420 EUR
PRSI45-421
Anti-CORO1A Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 60 kDa band observed in MOLT4 and Jurkat lysates (calculated MW of 51.0 kDa according to NP_009005.1). In transfected HEK293 transiently expressing CORO1A band of approximately 50 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended for use at 0.3-1 μg/mL.

Coronin 1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Coronin 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CORO1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR