Anti-COL11A1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-411EA 325 EUR
PRSI45-411
Anti-COL11A1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 28 kDa band in lysates of cell lines A431, A549, HEK293 after 0.5 μg/mL antibody staining. The 28 kDa band was successfully blocked by incubation with the immunizing peptide.

COL11A1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

COL11A1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: COL11A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR