Anti-IFT74 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-408EA 420 EUR
PRSI45-408
Anti-IFT74 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 70-75 kDa band observed in HEK293 lysate (calculated MW of 69.2 kDa according to AAK77221.1, but 51.4 kDa according to NP_079379.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Momeni et al, BMC Neurol. 2006 Dec 13;6:44.; PMID: 17166276). Recommended concentration: 0.3-1 μg/mL. Immunohistochemistry: In paraffin embedded human liver shows vesicular staining in cytoplasm of hepatocytes. Recommended concentration, 5-10 μg/mL.

CMG1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CMG1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IFT74
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR