Anti-CLCA1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-405EA 420 EUR
PRSI45-405
Anti-CLCA1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 26 kDa band in human duodenum, kidney and lung lysates after 0.3 μg/mL antibody staining.1. The 26 kDa band was successfully blocked by incubation with the immunizing peptide.

CLCA1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CLCA1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CLCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR