Anti-CHD7 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-396EA 420 EUR
PRSI45-396
Anti-CHD7 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 250 kDa and 48 kDa in rat spinal cord lysates after 1 μg/mL antibody staining. Both detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands).

CHD7 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CHD7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CHD7
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR