Anti-CDC42BPA Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-384EA 420 EUR
PRSI45-384
Anti-CDC42BPA Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 30 kDa and 40 kDa bands in human pancreas lysates at 1 μg/mL.3 and 196 kDa according to NP_003598.2. Immunohistochemistry: In paraffin embedded human cerebellum shows cytoplasm staining in Purkinje cells. Recommended concentration, 3-6 μg/mL.

Cdc42-binding kinase alpha antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Cdc42-binding kinase alpH a antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CDC42BPA
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR