Anti-FCGR2B Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-383EA 553 EUR
PRSI45-383
Anti-FCGR2B Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 34 kDa band observed in human myeloid erythroblast cell line K562 and human monocytic cell line U937 lysates (calculated MW of 34.0 kDa according to NP_003992.3;). Recommended concentration: 0.3-1 μg/mL. Immunohistochemistry: In paraffin embedded human liver shows membranous staining in endothelial cells between the hepatocytes. Similar results were obtained in human placenta. Recommended concentration, 3-6 μg/mL.

CD32 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CD32 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FCGR2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR