Anti-CTSK Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-366EA 420 EUR
PRSI45-366
Anti-CTSK Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 36 kDa band observed in human lung and human ovary lysates (calculated MW of 37.0 kDa according to NP_000387.1). Recommended concentration: 1-3 μg/mL.

Cathepsin K antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Cathepsin K antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CTSK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR