Anti-CNR1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-355EA 420 EUR
PRSI45-355
Anti-CNR1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 65 kDa band observed in rat brain lysates (calculated MW of 52.7 kDa according to rat NP_036916.1 and 52.9 kDa according to human NP_057167.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (PMID: 10762343). Recommended concentration: 0.1-0.3 μg/mL.

Cannabinoid Receptor 1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Cannabinoid Receptor 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CNR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR