Anti-CACNB4 Goat Polyclonal Antibody

PRSI45-351EA 546 EUR
PRSI45-351
Anti-CACNB4 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 60+55 kDa bands observed in human bone marrow lysates (calculated MW of 58.2 kDa according to NP_000717.2 and 54.7 kDa according to NP_001005747.1). Recommended concentration: 0.3-1 μg/mL.

CACNB4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CACNB4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CACNB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR