Anti-C14ORF68 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-346EA 553 EUR
PRSI45-346
Anti-C14ORF68 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 45 kDa band in human liver lysates after 0.3 μg/mL antibody staining.4 kDa according to NP_997000.2. The 45 kDa band was successfully blocked by incubation with the immunizing peptide.

C14orf68 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

C14orf68 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: C14ORF68
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR