Anti-CKB Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-339EA 420 EUR
PRSI45-339
Anti-CKB Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 45-48 kDa band observed in human brain lysates (calculated MW of 42.6 kDa according to NP_001814.2). Recommended concentration: 0.1-0.3 μg/mL.

Brain Creatine Kinase antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Brain Creatine Kinase antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR