Anti-BDKRB1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-338EA 420 EUR
PRSI45-338
Anti-BDKRB1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 35 kDa band observed in lysates of cell lines Daudi, K562 and U937 calculated MW of 40.5 kDa according to NP_000701.2). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. Recommended concentration: 2-4 μg/mL.

Bradykinin Receptor B1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Bradykinin Receptor B1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BDKRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR