Anti-BNIP1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-335EA 553 EUR
PRSI45-335
Anti-BNIP1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 28 kDa band observed in human kidney lysates (calculated MW of 26.1 kDa according to NP_001196.1; NP_053581.1; NP_053582.1; NP_053583.1). Recommended concentration: 0.5-1.5 μg/mL. an additional band of unknown identity was also consistently observed at 48 kDa . This band was successfully blocked by incubation with the immunizing peptide.

BNIP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

BNIP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BNIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR