Anti-BGN Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-329EA 420 EUR
PRSI45-329
Anti-BGN Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 38-40 kDa band observed in rat skin lysates (calculated MW of 41.7 kDa according to rat NP_058783.1 and human NP_001702.1 ). Recommended concentration: 0.1-0.3 μg/mL.

Biglycan Preproprotein antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Biglycan Preproprotein antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BGN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR