Anti-TRIM3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-327EA 420 EUR
PRSI45-327
Anti-TRIM3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 85 kDa band seen in mouse brain and human cerebellum lysate [Predicted MW of approximately 80.8 kDa according to NP_006449.2]. Recommended for use at 0.1-1 μg/mL.

BERP antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

BERP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRIM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR