Anti-BCAR3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-324EA 420 EUR
PRSI45-324
Anti-BCAR3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 110 kDa band observed in mouse embryonic kidney epithelial cell lysates (calculated MW of 92.6 kDa according to human NP_003558.1) and 92.3 kDa according to mouse NP_038895.1. Recommended concentration: 0.5-2 μg/mL.

BCAR3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

BCAR3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BCAR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR