Anti-BAG5 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-318EA 420 EUR
PRSI45-318
Anti-BAG5 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Approximately 55 + 48 kDa bands observed in nuclear HeLa cell lysate (calculated MW of 56.0 kDa according to NP_001015049.1 and of 51.2 kDa according to NP_004864.1; NP_001015048.1). Recommended concentration: 1-3 μg/mL.

BAG5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

BAG5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BAG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR