Anti-ACTL6A, ACTL6B Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-314EA 420 EUR
PRSI45-314
Anti-ACTL6A, ACTL6B Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 48 kDa band observed in Hela lysates (predicted MW of 49 kDa according to NP_057272, NP_004292). Recommended for use at 1-3 μg/mL.

BAF53A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

BAF53A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ACTL6A, ACTL6B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR