Anti-PPP2R5B Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-312EA 553 EUR
PRSI45-312
Anti-PPP2R5B Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Approximately 55 kDa band observed in human cerebellum lysates (calculated MW of 57.4 kDa according to NP_006235.1). Recommended concentration: 2-3 μg/mL.

B56 beta antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

B56 beta antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PPP2R5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR