Anti-EIF3M Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-311EA 420 EUR
PRSI45-311
Anti-EIF3M Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 38-40 kDa band observed in lysates of human lung carcinoma cell line A549 (calculated MW of 42.5 kDa according to NP_006351.2). Recommended concentration: 1-3 μg/mL. Immunohistochemistry: In paraffin embedded human testis shows strong staining of cytoplasm in select spermatocytes. Recommended concentration, 5-10 μg/mL.

B5 Receptor antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

B5 Receptor antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EIF3M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR