Anti-AXIN1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-310EA 420 EUR
PRSI45-310
Anti-AXIN1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 150 kDa and 22-24 kDa in human hippocampus lysates after 0.1 μg/mL antibody staining.6 kDa according to NP_003493.1 and 91.7 kDa according to NP_851393.1. The detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands). Immunohistochemistry: In paraffin embedded human adrenal gland shows membranous and fibrous staniing in and around secretory cells of the zona fasciculata in the cortex. Recommended concentration, 3-5 μg/mL.

AXIN1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

AXIN1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: AXIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR