Anti-AVPR1B Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-308EA 420 EUR
PRSI45-308
Anti-AVPR1B Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 170 kDa band observed in human brain (Substantia Nigra) lysates (calculated MW of 47.0 kDa according to NP_000698.1). The observed molecular weight corresponds to findings with EB08786, a product of different design reacting to the mouse protein. Recommended concentration: 0.05-0.2 μg/mL. Immunohistochemistry: In paraffin embedded human Pituitary shows bright cytoplasm staining of a few active secretory cells in the anterior pituitary. Recommended concentration, 5-10 μg/mL.

AVPR1B antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

AVPR1B antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: AVPR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR