Anti-ARL4A Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-286EA 420 EUR
PRSI45-286
Anti-ARL4A Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:1000. Western Blot: Approximately 23 kDa band seen in human testis lysate. Recommended for use at 1-3 μg/mL.

ARL4A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ARL4A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ARL4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR