Anti-ARF1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-277EA 553 EUR
PRSI45-277
Anti-ARF1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 18 kDa band observed in lysates of cell lines HEK293, A431 and HepG2 (calculated MW of 20.7 kDa according to NP_001019397.1; NP_001649.1; NP_001019398.1; NP_001019399.1). Recommended concentration: 1-3 μg/mL.

ARF1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ARF1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ARF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR