Anti-ANP32B Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-272EA 553 EUR
PRSI45-272
Anti-ANP32B Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: 30 kDa band observed in A431 and human lymph node lysates (predicted MW 29 kDa according to NP_006392). Recommended for use at 0.2-1 μg/mL.

APRIL antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

APRIL antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ANP32B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR