Anti-APOC1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-266EA 553 EUR
PRSI45-266
Anti-APOC1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Bands at 30 kDa and 37 kDa band in human liver lysates after 0.5 μg/mL antibody staining. Immunohistochemistry: In paraffin embedded human liver shows spotty staining in the cytoplasm of hepatocytes. Recommended concentration, 3-5 μg/mL.

APOC1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

APOC1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: APOC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR