Anti-ANXA10 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-255EA 420 EUR
PRSI45-255
Anti-ANXA10 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8000. Western Blot: At 1 μg/mL antibody banded at approximately 45-48 kDa in Hela, A431 and human kidney lysates. Immunohistochemistry: In paraffin embedded human pancreas shows staining of islet of Langerhans. Recommended concentration, 3-5 μg/mL.

Annexin A10 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Annexin A10 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ANXA10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR