Anti-ALOX5AP Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-241EA 420 EUR
PRSI45-241
Anti-ALOX5AP Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 48 kDa band in human brain and U937 lysates at 1 μg/mL. Further, this band was successfully blocked by incubation with the immunizing peptide. ALOX5AP antibody has been successfully used in WB assays with transfected HeLa cells (customer feedback, see Maier TJ et al., 2008).

ALOX5AP antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ALOX5AP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ALOX5AP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR