Anti-AIP Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI45-223EA 420 EUR
PRSI45-223
Anti-AIP Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 37 kDa band observed in NIH/3T3 lysate (calculated MW of 37.7 kDa according to human NP_003968.1 and 37.6 kDa according to mouse NP_057875.1). In transfected HEK293 transiently expressing ARA9 a band of approximately 42 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.02-0.06 μg/mL.

AIP antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

AIP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: AIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR